Autosampler configuration locks up Qtegra
Issue
The configuration with the autosampler after the iCAPPRO locks up Qtegra
Environment
- iCAPPRO
- Qtegra
Resolution
Make a new configuration with the autosampler plugin listed before the instrument configuration. Will need to make new lab books to run this configuration, if lab books had previously been made.