CID firmware error during installation
Issue
During the new install of an ICAPPRO all version the message "Camera firmware revision xxxxxx is newer than the supported version" appear in Log view in Qtegra dashboard.
Environment
- ICAPPRO
- ICAPPRO X
- ICAPPRO XP
- ICAPPRO XPS
Resolution
Install the latest version of Qtegra software
You can download a copy from Thermo Fisher Sharepoint or if you have no access to Sharepoint, contact Thermo Fisher Scientific.
Cause
Some ICAPPRO are supplied with old Qtegra software version and instrument have been tested or into CID it is loaded a newer version of Qtegra, when connecting the instrument the old version firmware installed in PC enters in conflict with loaded into camera
Attachment(s)
| File | Last Modified |
|---|---|
| log_view.png | May 31, 2023 |