Camera Cooling Interlock Warning Solved by Changing Configurations
Issue
Camera is not cooling. Wither the interlock is red or the camera is not reaching temperature.
Environment
- iCAP 6000/7000 Series
- iCAP PRO
Resolution
- System is found to not function due to either a camera temp interlock or camera not reaching temperature warning.
- System must be running Qtegra
- Change current configuration to Empty and leave for 1 minute
- Change the configuration back to the working configuration of choice.
- Once working configuration has loaded proceed with startup procedures and see if camera cools to designated temperature to clear interlock/warning.