Capillary tubing to Mixing Y disconnecting when pump is on high
Issue
When pump goes to high speed during uptake, capillary tubing coming nebulizer disconnects from the mixing Y causing leaks.
Environment
- iCAP 7000 Series
- iCAP PRO Series
Resolution
1. Remove the current piece of pump tubing on the mixing Y where nebulizer capillary tubing connects to
2. Cut off a small section of tubing from Orange/White pump tube and install on mixing Y
3. Cut the end of the capillary tubing going to mixing Y at a straight cut, no 45 degree
4. Using sand paper or glove to get a better grip on capillary tubing, push capillary tubing into pump tubing until it is about ¼” (approximately 6mm) of the way in
Analysis
The tubing does not disconnect when pump is at normal speed