Thermo Fisher Scientific
Determining Peristaltic Pump Model
This article discusses the replacement and repair of peristaltic pumps in the ICAP 7400 model, highlighting the importance of using compatible pump windings. It explains the differences between the "mini" style pump, with a smaller barrel and 3-stop/3-flag windings, and the regular pump, with a larger footprint and 2-stop/2-flag windings. Additionally, it mentions the ICAP 7600 model, which uses a mini style pump with an integrated vacuum valve interface, and provides information on the availabi
Issue
On the ICAP 7400 model there are two possible sizes for the peristaltic pump. If the pump needs to be replaced or repaired, it is important to replace with the same platform so that the pump windings you have will be compatible with the new pump. 
Environment
  • iCAP 6000 Series
  • iCAP 7000 Series
  • iCAP 7400
Resolution
The "mini" style peristaltic pump, pictured below,  is characterized by a smaller barrel and platens. It uses 3-stop/3-flag pump windings.
iCAP 7400 Peristaltic MiniPump - Complete                                 842315550761
The regular peristaltic pump, pictured below, is characterized by a larger footprint. It uses 2-stop/2-flag pump windings.
3-channel and 4-channel pump options are available, which references the number of platens/arms that are used. 
3 channel pump                                                                                                 430122821701
4 channel pump                                                                                                 430122821711
The ICAP 7600 uses a mini style pump but also includes the interface for the integrated vacuum valve, pictured below.
iCAP 7600 Peristaltic Pump Complete                                                  842315550771
For the regular peristaltic pump there is also a service kit available that can be ordered. This kit includes several wear items that can be replaced if the motor is still working and driving. A common reason to order this over the complete assembly is if some of the rollers on the barrel are stuck and unable to be lubricated but the entire barrel/pump head still spins. 
  • Was this article helpful?