Determining which tubing types are needed for the sample line uptake and the drain or waste line
Issue
How do I determine which tubing types are needed for the sample line uptake and the drain or waste line?
Environment
- iCAP 6000 Series
- iCAP 7000 Series
Resolution
The iCAP 6000 and 7000 Series use sample pump tubing to push sample into the nebulizer to the spray chamber to the center tube and into the plasma. The pump tubing has several different materials in use depending on the solvent used to dilute samples for analysis. The attachment will help determine which tubing types are needed for the sample line uptake and the drain or waste line.
Attachment(s)
| File | Last Modified |
|---|---|
| iCAP OES Sample Pump Tubing.pdf | August 02, 2022 |