Droplet Forming at the Tip of the Nebulizer
Issue
Droplet Forming at the Tip of the Nebulizer
Environment
- iCAP 6000 Series
- iCAP 7000 Series
- iCAP PRO
Resolution
- Remove the plastic spacer from the nebulizer and push the nebulizer fully into the spray chamber
- Flush the system for a couple of minutes with 10% Decon 90 solution or similar diluted detergent while plasma is on
- Flush system for a few minutes with DI water
- Remove nebulizer, install spacer and reinsert nebulizer fully into the spray chamber to the point where the spacer makes contact with the spray chamber.