Thermo Fisher Scientific
E2200 Plasma Ignition Failed 3kW DC power supply reported bad AC input
Issue
2200#: Plasma Ign Failed; 3kW DC power supply reported bad AC input.
Environment
  • iCAP 6000 Series
  • iCAP 7000 Series
Resolution
Replace the 3kW RF DC Power Supply 8423 155 50381
Analysis
  • Confirm input voltage to instrument.
  • Confirm there is no damaged wiring that supplies the DC PSU with power or communications.
  • If DC PSU LED is extinguished, replace DC PSU.
  • Was this article helpful?