Explanation of Forcing options
Issue
This article explains the difference between the options in the forcing section of the calibration.
Environment
- iCAP 6000 Series
- iCAP 7000 Series
- iCAP Q
- iCAP RQ
- iCAP TQ
Resolution
There are 3 options for forcing:
- None: Value y for x=0 is not manipulated
- Zero: The calibration curve will be forced to fulfill f(0)=0 which is equivalent to set a0 = 0.
- Blank: Defines the calibration curve f(x) with x=0 as:
- f(0)=iBLK Emission Line
- or F(0)=iAverage BLK Emission Line
depending on the current blank block mode
Explanation is from pg 13-8 Software manual BRE0015330, Rev B