Glass Expansion Trident Internal Standard Mixing Kit
Issue
Customer has with their iCAP a T-fitting that looks like PEEK material. Is this for internal standard (IS) mixer?
Environment
- iCAP 6500 Radial
- iCAP 6500 Dual
- iCAP 6300 Radial
- iCAP 6200 Dual
- iCAP 6300 Dual
Analysis
Customer has gotten the PEEK T-fitting directly from Glass Expansion. Our standard IS mixing kit, if used, is P/N 842312051551. The kit contains 1 complete assembly and extra pump windings. See the attached image of the mixing kit and application note.
The Glass Expansion - Trident Internal Standard Kit for HF solutions is P/N 60-808-1150 and contains:
| Part Number | Description | Part Number | Description |
| 60-808-1124 | HF Mixing Chamber/Tee | 60-808-1187 | Internal Standard Probe |
| UFT-EZT-16-50 | UniFit - EzyFit Sample Tube, 0.50mm ID (x1) | KK711E | Vitex Gripping Paper (PKT 6) |
| EZT-16-50 (x2) | EzyFit Sample Tube, 0.50mm ID (x1) | 70-803-0924 | Flared Connector |
| EZT-16-25 | EzyFit Internal Standard Tube, 0.25mm ID (x1) |
Resolution
Trident Dilution Factor Calculator
(available online at http://www.geicp.com/cgi-bin/site/wrapper.pl?c1=Products_accessories_internkit)
Sample pump tubing: orange/white, 0.64mm ID
Internal standard pump tubing: orange/blue, 0.25mm ID
Sample is diluted by 13.2%
Final concentration of sample is 0.868 times initial concentration
--------------------------------------------------------------------------------
Internal standard is diluted by 86.8%
Final concentration of internal standard is 0.132 times initial concentration
or Internal standard is diluted by a factor of 7.6
This calculation should be used as a guide only. Variations between pump tubes and roller pressures mean that the accuracy of the calculation cannot be guaranteed.
Attachment(s):
| File | Last Modified |
|---|---|
| GE Trident mixing kit.jpg | August 02, 2022 |
| Trident-CT_appnote.pdf | August 02, 2022 |