Humidity error displayed when Detector cooling begins during Get Ready
Issue
Humidity error displayed when Detector cooling begins during Get Ready.
Environment
iCAP PRO
Resolution
Multiple leaks repaired at Argon moisture trap followed by Engineer's purge for 2 hours allowed Detector cooling without humidity error.