Humidity in the camera is too high
Issue
Detector is failing to cool. An error is being displayed that indicates that the humidity in the camera is too high.
Environment
- iCAP PRO
Resolution
Enable the Engineering Purge
Analysis
The Engineering purge allows a higher level of gas flow through the Detector in order to potentially outgas any moisture present