Instrument Does Not Support Intelligent Dilution
Issue
Error: Instrument doesn't support Intelligent Dilution
Environment
- iCAP PRO
- Qtegra
Resolution
Inteligent Dilution is not supported in Qtegra 2.11. Upgrade Qtegra software to 2.14.
Analysis
ASX560 was upgraded with SDX autodilutor and this error was encoutered when trying to run a LabBook with autodilution enabled.
Cause