Thermo Fisher Scientific
Intensities are about half in Axial View
Issue
Significant reduction of sensitivity in Axial view but Radial view intensities are normal
Environment
  • iCAP PRO
Resolution
Power cycle the instrument
Analysis
A Fore Optics alignment was performed in Axial view.  However, it failed to bring the axial intensities up. Upon inspection of the Fore Optics autotune report, it was observed that the apex of the Optical Mirror 2 optimization was off the graph, meaning that the optimal position was not able to be found in the given Position range
For example, below is a good Optical Mirror 2 scan in which the apex of the curve is found within the Position range.
  • Was this article helpful?