Legacy ICP Internal Standard Mixing Kit
Issue
- Y connectors (p/n 13674600) are fragile and easy to damage.
- Kit tubing discolored (contaminated) at the Y; the fitting is partially collapsed where tubing connects;
- The fitting has is bent over where the tubing connects.
Environment
- iCAP 6500 Radial
- iCAP 6500 Dual
- iCAP 6300 Radial
- iCAP 6300 Dual
- iCAP 6200 Dual
Resolution
Customers can buy from Cole Parmer directly using their p/n K-30614-02 for nylon 1/16" y connector or K-30614-03 for polypropylene type. Each has 10 per pack.