Loss of intensities for Axial and Radial View
Issue
Intensities of wavelength (Y370 in this case) monitored in both axial and radial view decreased by an order of magnitude or more in both views.
Environment
- iCAP 6000 Series
- iCAP 7000 Series
Resolution
1) Perform Torch Alignment using 2ppm Zn solution such as Load Blank solution
2) Perform Autopeak adjust on affect wavelength(s)
3) Optimize number of center pixels using one of the knowledge articles below depending on whic software is being utlilized:
Determining the Number of Center Pixels in Qtegra for ICP-OES
Determining the Number of Center Pixels in iTEVA.pdf
Analysis
Y224 was also being monitored in axial view but did no suffer a loss of sensitivity.
Y370 may have been affected in this example but the principle would apply to any wavelength that has suffered a loss of sensitivity when others have not.