Manually Light Plasma-ICAP PRO
Issue
For plasma diagnostic purposes, it is sometimes necessary to manually light the plasma on the ICAP PRO via the Qtegra Service Application.
Environment
ICAP PRO
EAR99
Resolution
- With Service App open and connected to instrument, click on the Hardware Control tab
- Select PlasmaControl
- Set RF Power Supply Voltage in V to 35. Note the readback will not change, this is just the set point for now
- Select GasFlowControl
- Set Nebulizer Gas Flow to 0.00, Auxillary Gas Flow to 0.50, and Coolant Gas Flow to 12.00
- Select InstrumentControl
- Ensure that the water flow is higher than 2 l/min. RF Generator Cooling and Main Water Valve should be toggled on
- Toggle RF Power Supply Enable; check to make sure the readback matches the setpoint of 35 V
- Toggle RF Generator Enable
- Toggle RF Generator Ignite; note after the plasma lights this will toggle back off
- Check to see if plasma has lit and that the read-backs for RF Power Supply Voltage and Current are normal, and the plasma is stable
- To extinguish the plasma, toggle RF Power Supply Enable to OFF position.
Attachment(s)
| File | Last Modified |
|---|---|
| Disable.png | September 19, 2024 |
| Ignition.png | September 19, 2024 |
| Ignite.png | September 19, 2024 |
| RF gen enablecrop.png | September 19, 2024 |
| PS enabled-box.png | September 19, 2024 |
| Water flow.png | September 19, 2024 |
| IC select.png | September 19, 2024 |
| Gas flow set point.png | September 19, 2024 |
| Gas flow control page box.png | September 19, 2024 |
| Ps35 Box.png | September 19, 2024 |
| RF 0.png | September 19, 2024 |
| Set PS 35-PlasmaControl.png | September 19, 2024 |
| Set PS 35-Crop.png | September 19, 2024 |
| Set PS 35.png | September 19, 2024 |