Max amount of Acid ICP OES
Issue
How much acid can safely be used on the iCAP OES.
Environment
- iCAP 6000 Series
- iCAP 7000 Series
Resolution
iCAP OES can handle higher acid concentrations but the peri pump tubing will start to breakdown. So it is preferable to stay under 10% total acid. If it is necessary to use a higher acid concentration then Viton tubing can be purchased, but total acid should still be under 20%.