Thermo Fisher Scientific
Melting Torch aftermarket ESI Meinhard
Issue
Melting torch, aftermarket ESI/Meinhard
Environment
  • iCAP 6000 / 7000
  • Quartz torch
Resolution
  • Replace aftermarket torch with correct Thermo Fisher Scientific torch.
Analysis
  • Confirmed Proper Argon pressure and purity.
  • All O-rings in place.
  • Base and torch installed correctly.
  • Coil aligned as it should be.
  • Found an aftermarket torch installed.
Cause
Found that ESI / Meinhard torch had a manufacturing defect .  The orifice for the cool gas was improperly etched.  It would not allow the vortex of argon to occur, thus allowing the torch to melt.
  • Was this article helpful?