Moving a LabBook from One Computer to Another in Qtegra Software
This article provides a step-by-step guide to transferring a LabBook file from one computer to another within the Qtegra Intelligent Scientific Data Solution (ISDS) Software. The process involves locating the LabBook file on the source computer, copying it to an external storage device or network location, transferring it to the destination computer, and finally importing it into Qtegra on the destination computer. The article also includes optional internal steps to ensure compatibility and fil
Issue:
Users need to transfer a LabBook from one computer to another within the Qtegra software.
Environment:
- Product: Qtegra Intelligent Scientific Data Solution (ISDS) Software
Resolution:
(NOTE: Ensure both computers have compatible versions of Qtegra installed. Verify that the destination computer has the necessary permissions to access and modify the LabBook files.)
- Locate the LabBook File:
- The LabBook files are typically stored in the following file pathway: C:\ProgramData\Thermo\Qtegra\_Application Data\Workspace\LabBooks.
- Copy the LabBook File:
- Navigate to the file pathway mentioned above on the source computer.
- Copy the LabBook file (with the extension .imexp) to an external storage device or a network location accessible from the destination computer.
- Transfer the LabBook File:
- Connect the external storage device to the destination computer or access the network location where the LabBook file was copied.
- Paste the LabBook File:
- Paste the LabBook file into the same file pathway on the destination computer: C:\ProgramData\Thermo\Qtegra\_Application Data\Workspace\LabBooks.
- Open the LabBook in Qtegra:
- Open Qtegra on the destination computer.
- In the left-hand panel, select File Manager.
- Right-click on Lab Books and select "Import".
- Use the file manager to locate and select the LabBook file you transferred.
- The LabBook should now be visible and ready to use in Qtegra.
Analysis:
Cause: