Nebulizer Gas Auto Tune Failure
Issue
Optimization failed parameter not reached, desired value after 30 sec, parameter neb gas flow set value to set 0.85 last readback value 0.813.........
NOTE: set value and pressures may vary.
During the Nebulizer Auto Tune sequence, the Nebulizer Gas Flow readback fails reach the set value.
Environment
- iCAP PRO
Resolution
Neb pressure not reached due to neb clog or damage. Readback is not reaching set value. Replace nebulizer with new / unused unit.