Thermo Fisher Scientific
No Communiction with iSC-65
Issue
iSC-65 will not communicate within configuratoin in Qtegra
Environment
  • iCAP PRO
  • iCAP Qnova Series
Resolution
Use a pointed non-conducting tool or equivalent to press the Reset IP button for three seconds
Reset iSC-65.png
Analysis
Cause
  • Was this article helpful?