POP Window Cleaning
Issue
Correction of low intensities by cleaning the POP windows
Environment
- iCAP 6000
- iCAP 7000
Analysis
Low intensities on radial or axial view.
Cause
The POP window is dirty allowing minimal light to pass.
Attachment(s)
| File | Last Modified |
|---|---|
| Cleaning iCAP OES windows.pdf | March 02, 2023 |