Thermo Fisher Scientific
Peri pump not turning during lab book run
Originally submitted by kendall.norwood@thermofisher.com on 6/16/2025 8:09:49 PM UTC
Issue
The peri pump is on in standby, but won't turn when the lab book is running.
Environment
  • iCAP PRO
  • Qtegra
Resolution
The tune set settings were saved with the peri pump speed at 0. Open Tune Set editor Set the peri pump speed Click save Click activate on instrument.
Analysis
Cause