Thermo Fisher Scientific
Peristaltic Pump rebuild kit for ICP OES
Issue
Peristaltic Pump roller failure or head failure
Environment
  • iCAP 6000 series
  • iCAP 7000 series with large pump
Resolution
Install a peri pump rebuild kit, see image to follow:
Large peri pump repair kit and inst.jpg
Analysis
  • Individual roller seize up due to lack of lubrication over time.  Try a light machine oil to resolve.  Otherwise, replace roller drum assy.
  • Roller drum assy does not rotate, however motor is turning.  Replace the rubber motor coupling.
  • Rubber motor mounts have degraded or have broken.  
  • Pump tubing clamps are worn or failed.
Cause
Environmental factors (acidic fumes, humidity) as well as general wear and tear.
  • Was this article helpful?