Peristaltic Pump rebuild kit for iCAP Pro
Issue
Peristaltic Pump roller failure or head failure
Environment
iCAP Pro Series
Resolution
BRE0019644 iCAP PRO Peri Pump RepKit
The new style motor mounts (item H from picture above) look like this
Analysis
- Individual roller seize up due to lack of lubrication over time. Try a light machine oil to resolve. Otherwise, replace roller drum assy.
- Roller drum assy does not rotate, however motor is turning. Replace the rubber motor coupling.
- Rubber motor mounts have degraded or have broken.
- Pump tubing clamps are worn or failed.
Cause
Environmental factors (acidic fumes, humidity) as well as general wear and tear.