Peristaltic pump will not rotate
Issue
The peristaltic pump will not rotate after plasma ignition on the iCAP 6000/7000 series
Environment
iCAP 6000 Series
iCAP 7000 Series
Resolution
Power off the iCAP 6000/7000 for 20 minutes
Analysis
No peristaltic pump movement after plasma ignition
Cause
Communication issue