Thermo Fisher Scientific
Pseudo Saturation in Qtegra
Issue
Prerequisite: This procedure assumes that a Torch Alignment and Autopeak adjust has already been performed while troubleshooting this issue.
Sometimes a peak will appear to be saturated in the subarray by the presence of missing pixel(s) even though the amount of the element in the solution being analyzed should not cause the wavelength pertaining to that a element to be saturated (see Figure 1). This is what is commonly referred to as pseudo-saturation. If this occurs, please perform the following steps to try and correct it.
P1.png
clipboard_e1419363423fcdc5219230257838e2c05.png
 Figure 1: An apparent saturation of Na 589.592
Environment
 iCAP 6000, iCAP 7000
Resolution 
1)    Click on Intensities under Evaluation Results
clipboard_e85e23d57fb288e5e7b7c8307db84ebc1.png
Figure 2: Intensities
2)    Double-click on the result which contains a replicate which has saturated (the intensity value will be blue).
clipboard_e23ce456459201091b65274b136c98c0a.png
Figure 3: Intensities in blue
3)    Increase the center (‘C’) pixels to include the area under the peak (see Figure 4). Typically, this will involve increasing the center pixels by one on either side of the current grouping of center pixels. In the example in Figure 4, the center pixels have been increased from 3 to 5.
Note: occasionally you might have to increase the subarray width greater than the default of 12 and turn off a Left or Right background correction point.
clipboard_eccf145f3acc6281dd81302c5deedfe23.png
Figure 4: Center pixels were increased from 3 to 5 to totally encompass the peak
4)   Click on clipboard_e2f5e720193485fef21512b2606814533.png to save apply changes. to save apply changes.
5)    Aspirate a solution containing the element pertaining to the wavelength exhibiting pseudo-saturation and perform an Autopeak Adjust
Note: Do NOT perform an Autopeak Adjust on all wavelengths at the same time which are “saturating”. Instead, an Autopeak Adjust should be performed individually on each wavelength which is “saturating”.
6)    Using the same solution used to perform the autopeak adjust in step 5, run one repeat of an Unknown and then double click on the result in Intensities in order to view the Subarray
7)    Optimize the number of center pixels (see Figure 5)
clipboard_e682720a42eb5fc60f2efe2ea9840c129.png
Figure 5: Center pixels for Na 589.592 have been optimized.
8)    Click on clipboard_efaac6b7128509265ca91a43f44763a10.png  to save apply changes.
9)    Repeat steps 1-6 for any other wavelengths which are also exhibiting pseudo-saturation.
Analysis 
Missing pixels in subarray
Cause
Missing pixels in subarray
  • Was this article helpful?