Replacing the Pump Head on a Mini Peristaltic Pump
Issue
One or more of the rollers is binding up and therefore not moving freely.
Environment
- iCAP Q
- iCAP Qnova
- iCAP 7600
Resolution
Replace the pump head (P/N BRE0029335). This video will instruct you on how to do so.
Analysis
Cause