Request for location of Firmware Version
Issue:
How to find iCAP PRO Firmware version
Environment:
iCAP PRO
Resolution:
Customer can locate in LabBook
LabBook\iCAP PRO model\Instrument State\Readbacks: View Mode\User: Instrument Control Firmware Revision Readbacks
Other firmware information in Log Files:
C:\ ProgramData\Thermo\Qtegra\LogFiles