Resetting Qtegra with Empty Configureation
Issue
The get ready button can not be pushed and the plasma will not light.
Environment
- iCAP-OES
- iCAP-Q
- iCAP- RQ
Resolution
- Click on the dashboard to the left of the get ready button
- In the dialog box that has opened load the EMPTY Configuration.
- Once the EMPTY configuration has loaded, re-click the dashboard icon to the left of the get read button.
- In the dialog box that opens, click on the configuration you were trying to use and let it load.
- The get ready button show be able to be pushed, lighting the plasma.