Thermo Fisher Scientific
Sodium Emission is Not Centered in the Subarray
Issue
Sodium Emission is Not Centered in the Subarray. Performed Autopeak and while it passed still Sodium emission persists to be located at the top of the subarray and not centered in it.
Environment
  • iCAP PRO
Resolution
Performed Wavelengths Calibration in the Test Sets of the Service Software.
  • Was this article helpful?