Spectrometer Optimization Fails Due to Low Signal Intensity
Spectrometer optimization fails when signal intensity is insufficient for detection. This article provides step-by-step troubleshooting to isolate system state issues and sample introduction faults to restore normal operation.
Issue
Spectrometer optimization fails due to low signal intensity during startup or optimization routine.
Environment
- iCAP 6000 Series ICP-OES
- iCAP 7000 Series ICP-OES
- Qtegra Software
- iTEVA Software
Resolution
Method 1: Power Cycle the Instrument
- Close Qtegra software.
- Power off the instrument at the main breaker.
- Wait 30 minutes.
- Restart the computer. Do not open Qtegra.
- Turn on the instrument.
- Wait 5 minutes for initialization.
- Open Qtegra software.
- Select Get Ready to ignite the plasma and run spectrometer optimization.
Expected Result:
If optimization passes, the issue was related to system state.
Method 2: Isolate Sample Introduction System
- Block the sample introduction at the torch holder (e.g., temporarily seal to prevent sample flow).
- Run spectrometer optimization.
Expected Result:
- If optimization passes → Issue is caused by sample introduction components.
- If optimization fails → Further diagnostics required.
Component Replacement (if Method 2 passes)
Replace the following components:
- Torch holder O-rings
- Torch
- Center tube
Final Step
If spectrometer optimization continues to fail after these steps, perform a manual optics alignment.