Spray Chamber Flooding
Issue
The spray chamber is flooding
Environment
- iCAP 7000 Series
- iCAP PRO
Resolution
- Replace the drain peristaltic pump tubing and optimize the tension on the tubing
- Replace the capillary tubing that connects the spray chamber drain outlet to the drain peristalic pump tubing
- Verify that the end of the capillary tubing going from drain peristalic pump tubing to the waste vessel is not submerged in liquid. Also, it is a good idea to trim back a couple of inches of this tubing. If the issue still occurs, replace the piece of capillary tubing going from drain peristalic pump tubing to the waste vessel