Sprint Valve Errors Inhibiting LabBook from Running
Issue
Sprint valve errors (load failure command, etc.) occur which inhibit the LabBook from running even though the Sprint Valve is bypassed and not used
Environment
iCAP 7600
Resolution
Disabled the Sprint Valve by completing the following steps
1) Open Sprint Valve Configurator by opening the Windows Start Menu and then navigating to Sprint Valve Config
2) Click Connect to Sprint Valve
3) If the Enable Sprint Valve Operation box is checked,
uncheck it and click Save Configuration to Sprint Valve
If the Enable Sprint Valve Operation box is unchecked,
check it and click Save Configuration to Sprint Valve and proceed to step 4
Note: This may sound counterintuitive but there is a bug in the Sprint Valve Configurator software which enables its use in the beginning without showing the checkmark
4) Uncheck the Enable Sprint Valve Operation and click Save Configuratoin to Sprint Valve
Analysis
This only applies to version of Qtegra prior to 2.10 in which the Sprint Valve Configuartor application is used to control the Sprint Valve
Cause
Attachment(s)
| File | Last Modified |
|---|---|
| Unchecked_Enabled_Sprint_Valve_Operation.png | February 08, 2023 |
| Checked_Enabled_Sprint_Valve_Operation.png | February 08, 2023 |
| Spring Valve Config.png | August 02, 2022 |
| Connect to sprint Valve.png | August 02, 2022 |
| Save Configuration to Sprint Valve.png | August 02, 2022 |
| Checked Enabled Sprint Valve Operation.png | August 02, 2022 |
| Unchecked Enabled Sprint Valve Operation.png | August 02, 2022 |