Torch box Interlock Error
Issue
Torch box interlock showing up red
Environment
- iCAP 7000 series
- iCAP 6000 series
Resolution
- Remove the torch
- Verify that the red knob on the side of the torch holder is present, see black rectangle in picture below.
- If this is missing then you need a new torch holder
- If it is there, then try and to re-install the torch and make sure that it clicks into place.
- If the interlock is still red, then open and re-close the torch box door
- If the issue persists, contact Technical Support. Please have your serial number ready.