Thermo Fisher Scientific
Water flow sensor red, Detector Temp interlock not green
Issue
Water Flow sensor red, detector temperature red/blue
Environment
iCAP 7600
Resolution
  1. With plasma off, turn off the chiller
  2. Reverse water lines in the back of the chiller
  3. Run for 5 minutes
  4. Turn off chiller
  5. Replace chiller lines into the correct positions
  6. Turn on chiller
  7. Monitor interlocks
Analysis
Cause