Thermo Fisher Scientific
iCAP 6000 7000 Errors and Causes
Issue
An issue occurred  such as the plasma not igniting or going out and in iTEVA or Qtegra an error code was triggered. What does the error code refer to? And what potential corrective action are there? The list below is not comprehensive by any means but will list a lot of the error codes and corrective actions for some of them.
In iTEVA the error code will be displayed in the Journal and in Qtegra it will be displayed in the Log View. If you can't find the error in the Log View look in the most recent log file.
The Log file can be retrieved as follows:
1) Navigate to: Path:          C:\ProgramData\Thermo\Qtegra\LogFiles
2) Sort the contents of the LogFiles directory with the newest files at top of list by clicking on the "Date Modfied" column
3) Locate the most recent log file of the type:
File Name:  log-20140922-074323638.log  (format:  log-yyyymmdd-nnnnnnnnn.log)
Date & Time inside file: 2014-09-22T09:53:30.8412240-05:00|
                                       (Date)        (Time)
Note: If there is no general log file for the day in which the event occurred, please copy the general log file directly preceding the event in chronological order.
Environment
  • iCAP 6000 Series
  • iCAP 7000 Series
Resolution
iCAP - 6000/7000 ICP /  Errors List /  
Category Type Error Codes / Description Corrective Action
Initialization Firmware initialization errors 101#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 102#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 104#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 105#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 106#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 107#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 108#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 109#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 110#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 111#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 112#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 113#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 114#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 115#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 116#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware initialization errors 117#: Fatal Firmware error.  If the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 200#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 201#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 202#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 203#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 204#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 205#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 206#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 207#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 208#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 209#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 210#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 211#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Initialization Firmware configuration initialization errors 212#: Firmware failed to initialise successfully due to incorrect or missing configuration files.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.   
Firmware Firmware error handler errors 300#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 301#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 400#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 401#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 402#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 500#: Internal Firmware error - Camera driver not initialised, check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 501#: Internal Firmware error - Camera driver: Configuration appears to contain invalid parameters for this camera, check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 502#: Internal firmware error - Camera driver: If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 503#: Internal firmware error - Camera driver: If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 504#: Internal firmware error - Camera driver: If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 505#: Internal Firmware error - Camera driver: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 506#: Internal Firmware error - Camera driver: If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 507#: Internal Firmware error - Camera driver: If the condition persists please contact customer support with this error code.
See Ver 5.2 - iCAP Diagnostics Manual.pdf
Firmware Firmware error handler errors 508#: Internal Firmware error - Camera driver: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 509#: Internal Firmware error - Camera driver: Error programming FPGA. If the condition persists please contact customer support with this error code. See Ver 5.2 - iCAP Diagnostics Manual.pdf
Firmware Firmware error handler errors 510#: Internal Firmware error - Camera driver: Error Installing DMA ISR. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 511#: Internal Firmware error - Camera driver: PCI comms loop-back test failed during initialisation. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 512#: Internal Firmware error - Camera driver: Device not found on the PCI bus.  If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 513#: Internal Firmware error - Camera driver: Timeout while polling FPGA. If the condition persists please contact customer support with this error code. See Ver 5.2 - iCAP Diagnostics Manual.pdf
Firmware Firmware error handler errors 514#: Internal Firmware error - Camera driver: Invalid shutter timer value from FPGA, check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 516#: Internal Firmware error - Camera driver: Invalid pixel locations detected in camera driver, check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 517#: Autobias failure due to timeout - check flash up LED at rear of polychromator. If the condition persists please contact customer support with this error code. See Ver 5.2 - iCAP Diagnostics Manual.pdf
Firmware Firmware error handler errors 600#: Internal Firmware error - CID86Process: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 601#: Internal Firmware error - CID86Process: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 602#: Internal Firmware error - CID86Process: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 603#: Internal Firmware error - CID86Process: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 604#: Internal Firmware error - CID86Process: Check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 605#: Internal Firmware error - CID86Process: Invalid pixel locations detected, check firmware log for details. If the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 606#: Internal Firmware error - CID process - Invalid configuration settings detected.  The instrument may not be correctly configured, if the condition persists please contact customer support with this error code.  
Firmware Firmware error handler errors 607#: Internal Firmware error - CID86Process: Invalid memory manager memory pointers, check firmware log for details.  
Firmware Config commands 1000#: File Download to instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1001#: File Download to instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1002#: File Download to instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1003#: File Download to instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1004#: File Download to instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1005#: File Upload from instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1006#: File Upload from instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1007#: File Clear all on instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1008#: File Clear all on instrument failed. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1009#: Writing Secondary Sample introduction settings error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1010#: Sample introduction settings not found. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1011#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1012#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1013#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1014#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1015#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1016#: Secondary sample introduction error: Invalid pulse transition. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1017#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1018#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1019#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1020#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1021#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1022#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1023#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1024#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1025#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1026#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1027#: Firmware configuration setting error. If the condition persists please contact customer support with this error code.  
Firmware Config commands 1028#: Firmware configuration setting error: Check firmware log for details, if the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1101#: Analysis error (invalid number of subexposures) - exposure aborted  
Analysis OES exposure commands 1102#: Analysis error (invalid view type for this instrument) - exposure aborted  
Analysis OES exposure commands 1103#: Analysis error (invalid slit type for this instrument) - exposure aborted  
Analysis OES exposure commands 1104#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1105#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1106#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1107#: Analysis error (invalid parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1108#: Analysis error (invalid parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1109#: Analysis error (invalid exposure type for this instrument) - exposure aborted  
Analysis OES exposure commands 1110#: Analysis error (invalid number of repeats) - exposure aborted  
Analysis OES exposure commands 1111#: Analysis error (invalid acquisition mode for this instrument) - exposure aborted  
Analysis OES exposure commands 1112#: Analysis error (invalid parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1113#: Analysis error (invalid parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1115#: Analysis error (invalid number of lines per subexposure) - exposure aborted  
Analysis OES exposure commands 1116#: Analysis error (number of lines exceeded) - exposure aborted  
Analysis OES exposure commands 1117#: Analysis error (invalid exposure time) - exposure aborted  
Analysis OES exposure commands 1118#: Analysis error (invalid exposure time) - exposure aborted  
Analysis OES exposure commands 1119#: Analysis error (invalid time increment) - exposure aborted  
Analysis OES exposure commands 1120#: Analysis error (invalid number of subexposures) - exposure aborted  
Analysis OES exposure commands 1121#: Analysis error (number of lines exceeded) - exposure aborted  
Analysis OES exposure commands 1122#: Analysis error (invalid exposure time) - exposure aborted  
Analysis OES exposure commands 1123#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1124#: Analysis error (invalid exposure time) - exposure aborted  
Analysis OES exposure commands 1125#: Analysis error (unsupported exposure parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1126#: Analysis error (unsupported exposure parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1127#: Analysis error (unsupported exposure parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1128#: Analysis error (unsupported exposure parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1129#: Analysis error (unsupported exposure parameter) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1130#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1131#: Analysis error (invalid subarray coordinates) - exposure aborted  
Analysis OES exposure commands 1133#: Plasma went out during the exposure - exposure aborted  
Analysis OES exposure commands 1134#: Analysis error (invalid LED state for exposure) - exposure aborted  
Analysis OES exposure commands 1135#: Analysis error (invalid shutter position for exposure) - exposure aborted  
Analysis OES exposure commands 1136#: Analysis error (invalid number of time slices) - exposure aborted  
Analysis OES exposure commands 1137#: Analysis error (invalid viewing height) - exposure aborted  
Analysis OES exposure commands 1138#: Invalid torch compartment interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1139#: Invalid purge gas pressure interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1140#: Invalid torch gas pressure interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1141#: Invalid water flow interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1142#: Invalid drain flow sensor interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1143#: Invalid exhaust interlock detected during exposure - exposure aborted  
Analysis OES exposure commands 1200#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1201#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1202#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1203#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1204#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1205#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis OES exposure commands 1206#: Analysis cancelled by user - exposure aborted  
Analysis OES exposure commands 1207#: Analysis error (exposure data too large) - exposure aborted  
Analysis OES exposure commands 1208#: Analysis error (invalid argument, check instrument configuration and firmware log for details) - exposure aborted. If the condition persists please contact customer support with this error code.   
Analysis Generic run exposure system errors 1209#: Analysis error - exposure aborted, check firmware log for details. If the condition persists please contact customer support with this error code.  
Analysis Generic run exposure system errors 1210#: Analysis error - exposure aborted, check firmware log for details. If the condition persists please contact customer support with this error code.  
Analysis Generic run exposure system errors 1211#: Analysis error - (CID all zero's - Check detector and detector configuration) exposure aborted. If the condition persists please contact customer support with this error code.
Check to see if system can run FF image. The instrument can be powered down.  Remove camera RS 232 cables and reconnect them. Power up and check for analysis functions.
Analysis Generic run exposure system errors 1212#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis Generic run exposure system errors 1213#: Analysis error (return data buffer full) - exposure aborted. If the condition persists please contact customer support with this error code.  
Analysis Generic run exposure system errors 1214#: Analysis error (invalid sample introduction method requested of instrument).    
Analysis Generic run exposure system errors 1215#: Analysis error - exposure aborted. If the condition persists please contact customer support with this error code.  
    1216#: RF Power could not maintain requested power - If the actual RF power is out of range (+/- 20 W) for more than 20 seconds you will get error E1216 Is power reading back with set point?  If so, Check Argon Pressure, Exhaust Flow, coil position, Nebulizer; basically anything which could be causing plasma conditions to change.
    1217#: Auxillary Gas Flow - Count not maintain requested value.  
    1218#: Nebulizer Gas Flow Error - Could not maintain requested value.   
    1219#: Coolant Gas Flow - Could not maintain requested value  
Analysis Generic run exposure system errors 1250#: Detector not ready to process command - operation aborted. If the condition persists please contact customer support with this error code.  
Analysis Generic run exposure system errors 1251#: Firmware not ready to process command - operation aborted. If the condition persists please contact customer support with this error code. Goto Empty Configuraon in Qtegra, Open up iTEVA and handshake with instrument through iTEVA, Close iTEVA, and then Go to active configuraon in Qtegra. Make sure camera config is correct
Miscellaneous Misc commands 1700#: Firmware download failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1701#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1702#: Firmware retrieval of error logs failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1703#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1704#: Firmware download IO Board firmware failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1705#: Firmware download RF Unit firmware failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1706#: Firmware download TSP FPGA firmware failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1707#: CIDTEST Firmware download failed. If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1708#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1709#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1710#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1711#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
Miscellaneous Misc commands 1712#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
CID CID commands 1800#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
CID CID commands 1810#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
CID CID commands 1811#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
CID CID commands 1812#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
CID CID commands 1813#: Internal firmware error.  If the condition persists please contact customer support with this error code.  
I/O I/O commands 2000#: Firmware serial port could not be initialised, instrument must be restarted. If the condition persists please contact customer support with this error code.  
I/O I/O commands 2002#: IO communications error (RX data error), check firmware log for details. If the condition persists please contact customer support with this error code.  
I/O I/O commands 2003#: IO Board firmware download failed, check firmware log for details. If the condition persists please contact customer support with this error code.  
I/O I/O commands 2005#: IO Board not in a state to process commands. If the condition persists please contact customer support with this error code.  
I/O I/O commands 2006#: RF firmware download failed, check firmware log for details. If the condition persists please contact customer support with this error code.  
I/O I/O commands 2007#: RF firmware download error, please restart the instrument to reset instrument communications. If the condition persists please contact customer support with this error code.   
RF generated by RF commands 2011#: Pump speed invalid:  speed unchanged  
RF generated by RF commands 2012#: RF power invalid:  power unchanged  
RF generated by RF commands 2013#: Auxiliary Gas flow invalid:  flow unchanged  
RF generated by RF commands 2014#: Nebuliser Gas flow invalid:  flow unchanged  
RF generated by RF commands 2015#: Coolant Gas flow invalid:  flow unchanged  
RF generated by RF commands 2016#: Additional Gas flow invalid: flow unchanged  
RF generated by RF commands 2017#: Failed to set TE Power  
RF generated by RF commands 2018#: Failed to read TE Power  
RF generated by RF commands 2019#: Failed to read TE Status  
RF generated by RF commands 2020#: Failed to set Tank power  
RF generated by RF commands 2021#: Failed to read TE Power  
RF generated by RF commands 2022#: Failed to read Gas Solenoid Status  
RF generated by RF commands 2023#: Failed to set Gas Solenoid  
RF generated by RF commands 2024#: Failed to read water temperature  
RF generated by RF commands 2025#: Failed to read interlock status  
RF generated by RF commands 2026#: Failed to set pump speed  
RF generated by RF commands 2027#: Failed to read pump speed  
RF generated by RF commands 2028#: Failed to set pump direction  
RF generated by RF commands 2029#: Failed to read pump direction  
RF generated by RF commands 2030#: Failed to read nebuliser gas pressure  
RF generated by RF commands 2031#: Failed to set RF power  
RF generated by RF commands 2032#: Failed to read RF power  
RF generated by RF commands 2033#: Failed to read RF status  
RF generated by RF commands 2034#: Failed to set auxiliary gas flow  
RF generated by RF commands 2035#: Failed to read auxiliary gas flow  
RF generated by RF commands 2036#: Failed to set nebuliser gas flow  
RF generated by RF commands 2037#: Failed to read nebuliser gas flow  
RF generated by RF commands 2038#: Failed to set coolant gas flow  
RF generated by RF commands 2039#: Failed to read coolant gas flow  
RF generated by RF commands 2040#: Failed to set additional gas flow  
RF generated by RF commands 2041#: Failed to read additional gas flow  
RF generated by RF commands 2042#: Failed to set motor M1 position  
RF generated by RF commands 2043#: Failed to set motor M2Slit position  
RF generated by RF commands 2044#: Failed to set motor M2View position  
RF generated by RF commands 2045#: Failed to read motor M1 position  
RF generated by RF commands 2046#: Failed to read motor M2 position  
RF generated by RF commands 2047#: Failed to set SSEA triggers   
RF generated by RF commands 2048#: Failed to read SSEA triggers  
RF generated by RF commands 2049#: Failed to read IO Board status  
RF generated by RF commands 2050#: Failed to read IO Board version information  
RF generated by RF commands 2051#: Failed to read RF version information  
RF generated by RF commands 2052#: Failed to set view  
RF generated by RF commands 2053#: Failed to set slit  
RF generated by RF commands 2054#: Failed to set plasma  
RF generated by RF commands 2055#: Failed to abort IO operation  
RF generated by RF commands 2056#: Slit specified invalid for this instrument.  
RF generated by RF commands 2057#: View specified invalid for this instrument.  
RF generated by RF commands 2058#: Motor specified invalid for this instrument.  
RF generated by RF commands 2059#: Invalid Motor M1 position requested.  
RF generated by RF commands 2060#: Invalid Motor M2SLIT position requested Misaligned fore optics. Check mirrors and perform foreoptic alignment
RF generated by RF commands 2061#: Invalid Motor M2VIEW position requested  
RF generated by RF commands 2062#: Failed to set plasma purge delay  
RF generated by RF commands 2063#: Invalid plasma purge delay specified requested  
RF generated by RF commands 2064#: Failed to set DAC value  
RF generated by RF commands 2065#: Failed to get ADC value  
RF generated by RF commands 2067#: Failed to initialise motors. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2068#: Unsupported command for this instrument type (Basic Gasbox) - Command failed. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2069#: Failed to find M2 View ( Major ) motor end-stop, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2070#: Failed to find M2 Slit ( Minor ) motor end-stop, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2071#: Plasma ignition failed, reason for failure unknown.    
RF generated by RF commands 2072#: Failed to read RF FET Temp  
RF generated by RF commands 2073#: Failed to read debug bytes   
RF generated by RF commands 2074#: Slit Mask motor position specified invalid  
RF generated by RF commands 2075#: Failed to set slit mask motor position  
RF generated by RF commands 2076#: Firmware failed to get a valid IO Board version number, system not initialised.  Restart instrument and if the condition persists please contact customer support with this error code.  
RF generated by RF commands 2077#: Firmware failed to get a valid RF version number, system not initialised.  Restart instrument and if the condition persists please contact customer support with this error code.  
RF generated by RF commands 2078#: Failed during wait for ETV Ready.  
RF generated by RF commands 2079#: Failed to set Drain Flow Sensor state.  If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2080#: One of the fore-optics motors timed-out before reaching desired position, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2081#: The M2 View motor timed-out before reaching the desired position, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2082#: The M2 Slit motor timed-out before reaching the desired position, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2083#: The M1 motor timed-out before reaching the desired position, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2084#: The Slit Mask motor timed-out before reaching the desired position, fore-optics motor positions are now invalid. If the condition persists please contact customer support with this error code.  
    2198#: Water flow error  
RF generated by RF commands 2199#: Failed to extinguish plasma.  
RF generated by RF commands 2071#: Plasma ignition failed, reason for failure unknown.  
    2200#: Plasma ignition failed, 3kw Power supply reported bad AC input  
RF generated by RF commands 2201#: Plasma ignition failed, RF status failed. If the condition persists please contact customer support with this error code.  
RF generated by RF commands 2202#: Plasma ignition failed, RF reported bad supply power output failure  
RF generated by RF commands 2203#: Plasma ignition failed, RF reported bad argon flow interlock failure  
RF generated by RF commands 2204#: Plasma ignition failed, RF reported bad water flow interlock failure  
RF generated by RF commands 2205#: Plasma ignition failed, RF reported over temperature failure Could be RF Board or RF Interface board. This error can also be caused by low water flow
RF generated by RF commands 2206#: Plasma ignition failed, RF reported over current failure   
    2208#: Plasma ignition failed; RF seems OK. Check your sample introduction system as described in iTEVA Help. If the condition persists please contact customer support with this error code.  
    2209#: Plasma ignition failed; low ignition current. If the condition persists please contact customer support with this error code  
Interlocks instrument instrument interlocks and not RF interlocks 2210#: Plasma ignition failed, the torch compartment interlock is bad  
Interlocks instrument instrument interlocks and not RF interlocks 2211#: Plasma ignition failed, the purge gas interlock is bad  
Interlocks instrument instrument interlocks and not RF interlocks 2212#: Plasma ignition failed, the plasma gas interlock is bad  
Interlocks instrument instrument interlocks and not RF interlocks 2213#: Plasma ignition failed, the water flow interlock is bad  
Interlocks instrument instrument interlocks and not RF interlocks 2215#: Plasma ignition failed, the exhaust flow interlock is bad  
I/O IO Board communications errors 2900#: IO Board communications error - TX data write error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2901#: IO Board communications error - wait for port timeout. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2902#: IO Board communications error - Mutex release error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2903#: IO Board communications error - RX data timeout or incorrect number of bytes received. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2904#: IO Board communications error - IO Board detected protocol compare error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2905#: IO Board communications error - IO Board detected protocol frame error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2906#: IO Board communications error - IO Board detected parity error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2907#: IO Board communications error - IO Board all zeros response. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2908#: IO Board communications error - IO Board detected undefined protocol error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2909#: IO Board communications error - IO Board detected parameter error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2910#: IO Board communications error - IO Board detected message sequencing error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2911#: IO Board communications error - IO Board detected unknown message error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2912#: IO Board communications error - IO Board detected undefined message error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2913#: IO Board communications error - RX data protocol mismatch error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2914#: IO Board communications error - RX data re-program protocol mismatch error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2915#: IO Board communications error - IO Board comms time-out error. If the condition persists please contact customer support with this error code.  
I/O IO Board communications errors 2916#: IO Board communications error - IO Board comms overrun error. If the condition persists please contact customer support with this error code.  
RF RF commands 3001#: RF firmware download error (RF failed to enter program mode) - download failed. If the condition persists please contact customer support with this error code.  
RF RF commands 3002#: RF firmware download error (write data failed) - download failed. If the condition persists please contact customer support with this error code.  
RF RF commands 3003#: RF firmware download error (RF failed to exit program mode) - download failed.  Please restart the instrument to reset instrument communications. If the condition persists please contact customer support with this error code.  
RF RF commands 3004#: RF firmware download error (checksum get failed) - download failed. If the condition persists please contact customer support with this error code.  
RF RF commands 3005#: RF firmware download error (checksum check failed) - download failed. If the condition persists please contact customer support with this error code.  
RF RF commands 3006#: RF firmware download error (firmware write failed) - download failed. If the condition persists please contact customer support with this error code.  
RF RF commands 3010#: RF firmware download error (RF failed to restart) - download failed.  Please restart the instrument to reset instrument communications. If the condition persists please contact customer support with this error code./  
    3542#: Not showing up in the subarray Wavecal corrupted. Ran a new wavecal
    9000#: Plasma ignition failed; reason unknown  
Plasma Reasons for the plasma going out 9002#: Plasma went out unexpectedly, reason unknown.  
Plasma Reasons for the plasma going out 9002#: Plasma went out unexpectedly, RF reported bad supply power output failure.  
Plasma Reasons for the plasma going out 9003#: Plasma went out unexpectedly, RF reported bad argon flow interlock failure.  
Plasma Reasons for the plasma going out 9004#: Plasma went out unexpectedly, RF reported bad water flow interlock failure.  
Plasma Reasons for the plasma going out 9005#: Plasma went out unexpectedly, RF reported over temperature failure.  
Plasma Reasons for the plasma going out 9006#: Plasma went out unexpectedly, RF reported over current failure.   
Interlocks instrument instrument interlocks and not RF interlocks 9010#: Plasma went out unexpectedly, the torch compartment interlock is bad.  
Interlocks instrument instrument interlocks and not RF interlocks 9011#: Plasma went out unexpectedly, the purge gas interlock is bad.  
Interlocks instrument instrument interlocks and not RF interlocks 9012#: Plasma went out unexpectedly, the plasma gas interlock is bad.  
Interlocks instrument instrument interlocks and not RF interlocks 9013#: Plasma went out unexpectedly, the water flow interlock is bad.  
Interlocks instrument instrument interlocks and not RF interlocks 9014#: Plasma went out unexpectedly, the drain flow interlock is bad.  
Interlocks instrument instrument interlocks and not RF interlocks 9015#: Plasma went out unexpectedly, the exhaust flow interlock is bad.  
    33015:   
    33017: Spectrometer Optimization failed, peak too weak or not found  
    33018: Unable to complete torch alignment. Can’t locate edges of plasma image. Carryout foreoptic prealignment again.  
    33024: Debug Spectrometer Optimize failed  
    33100: Debug IIO Internal software error. General error in comms, the command to take a sample failed for some unknown reason This generally can be ignored.
    33101: Debug Invalid command response code returned Bypass internal comm cable
    33203: Failed to set plasma conditions  
    33204: Warning - Failure to load Wavelength Check file, calcheck bin  
CID Camera error  33207#. Detector un-initialised See Diagnostics Manual v2.2 - Instrument Error Codes
    33207: Unknown detector error has occurred  
I/O I/O commands 33208#. I/O & RF Communications Errors See Diagnostics Manual v2.2 - RF and I/O Problems
I/O I/O commands 33209#. I/O & RF Communications Errors See Diagnostics Manual v2.2 - RF and I/O Problems
    33210# 33210#  The configuration files(ini files) were incomplete or missing Load empty configuration in Qtegra and leave program.  Or reboot PC and Power off then on iCAP. Open iTEVA and allow the instrument to initialize.  Once done. Start up plasma.  Once you have confirmed plasma control and no problem, switch off plasma and close iTEVA. Open Qtegra and you should be okay. The INI files will be re-established from camera unless you have put in a new camera.  If you have put in a new camera, one must run all the alignment and run the Wavelength calibration in the manufacturing to establish these missing files
Firmware Firmware error 33211#. The firmware recently rebooted due to a non recoverable error.  The firmware Error log will be downloaded See Diagnostics Manual v2.2 - Instrument Error Codes
I/O I/O commands 33212#. An I/O board communications error has occurred.  Ensure the controller board is powered and connected correctly, if the condition persists please contact customer support. See Diagnostics Manual v2.2 - RF and I/O Problems
RF RF commands 33213# - An RF generator communications error has occurred. Ensure the RF generator is powered and connected correctly, if the condition persists please contact customer support RF power supply fan cycles continuously due to comm loop. Replace RF Interface board. See Diagnostics Manual v2.2 - RF and I/O Problems
    33214: A communications timeout has occurred during instrument initialization I would have the customer to wait for 3 minutes after the instrument has been switch on before opening Qtegra. This would allow the RF gen to go thru its initialization phase, which all of them do. After 3 minutes, then open the software and tell system to turn on.
Analysis Cetac Autosampler 33450# - Cetac Autosampler error, No Response to Command  
    33495# plasma went out unexpectectedly;  RF Gen temp rose to exceed 27 deg; water flow blockage or pump head deteriated; check circulator pressure for drop when plasma starts; no drop then blockage or pump flow failure;
    33535 /*Error Updating Method with standard signals and coefficients*/  
    33536  /*"#33536#: Checksum invalid in %s table. Record ID %s. The database may have been edited outside iTEVA. Data invalid - this record should be deleted."*/  
    33537  /*"#33537#: Failed to write Audit Trail entry to database."*/  
    33538  /*"#33538#: Failed to write Signature to database."*/  
    33539# Error Updating Method with standard: Standard saturated.
The reason for this is because the band of one of the analyzed elements is overlapping with something else with higher concentration..
    33540 ERROR_UPDATING_METHOD_WITH_STANDARD_SUBARRAY  
    33541 ERROR_UPDATING_METHOD_WITH_STANDARD_PLASMA  
    33542 ERROR_UPDATING_METHOD_WITH_STANDARD_PEAK_OFF_CHIP  
ce this text to add the Resolution.
Analysis
Cause