iCAP PRO- Loss of Sensitivity
Issue
Loss of sensitivity
Environment
- iCAP PRO
Resolution
Inspect the periscope mirror, if noticeably degraded, then replace.
Analysis
Cause
The periscope mirror is exposed to UV radiation the entire time that the plasma is on and will degrade at a much quicker rate than the mirrors in the optical tank that are sheilded.