iCAP PRO RF Coil Replacement
Issue
RF Coil needs to be replaced due to damage or other causes.
Environment
- iCAP PRO (All models)
Resolution
Replacing the RF Coil BRE0019591
- Make sure the chiller is off
- Open the torch box door
- If the instrument is a duo, remove the axial cone
- Remove the inner torch box, by removing the 3 screws, 2.5mm allen screws
- Remove the radial cone, by twisting counter clockwise
- Locate the 3mm allen screws that hold on the load coil on the right side in the teflon block. *Place a paper towel or paper over the gap in order to not lose any screws or tools.
- Unscrew until the RF coil is able to be wiggled free from the teflon block. (NOTE: The screws will come all the way out if not careful.)
- Remove coil from torch box.
- Replace new coil in the teflon block. The connections are keyed so it fits only one direction. Hold coil tight against teflon block to seat o-rings as you tighten the screws.
- Replace radial cone
- Replace the inner torch box
- Replace the axial cone if applicable
- Replace sample introduction and carry on.