iCAP Pro Plasma TV Camera will not Communicate
Issue
The iCAP Pro Plasma TV will not display.
Environment
- iCAP Pro Duo
- iCAP Pro Radial
Resolution
1) Open the command prompt and type the command arp -a
2) The Interface listed with the IP Address 192.168.1.100 is connection to the iCAP Pro
3) Locate a dynamic IP Address listed under Interface: 192.168.1.100 that starts with 192.168.X.XX other than 192.168.0.60 which is the Instrument Control Board and 192.168.1.2 which is the CID Detector.
For example, in the picture below, 192.168.0.88 is the Plasma TV IP address.
4) If there are only two dynamic addresses (192.168.1.2 and 192.168.0.60) mentioned, ping the following IPs and rerun "arp -a" 192.168.0.88, 192.168.1.88, 192.168.0.80, 192.168.1.80.
5) You should be able to see a third dynamic IP address now when running the "arp -a" command and will have to add this to the configuration in configurator.
6) In Qtegra Dashboard, Load an Empty Configuration (no plugins present in configuration)
7) Open Configurator
8) Click on Experiment Configurator
9) Locate the configuration that is normal used to operate the instrument and click on white triangle to expand the configuration to reveal the plugins contained within the configuration
10) Right-click on the iCAP PRO Series plugin and click Edit Settings
11) Change the IP Address under Plasma TV Settings to the value noted in step 3
Note: if the Plasma TV IP address is set up properly but it still won't communicate then you may need to re-initialize the configuration by going to an Empty configuration and re-loading the configuration used to for analysis. Consult this knowledge article: Qtegra for ICP-MS and ICP-OES Software Issues for more info on how to re-initialize the configuration.
12) Click Save
13) Load the Configuration that changes were just made to
Cause
The camera IP address is modified by Windows and it is necessary to use the command prompt to search for the IP address.
Attachment(s)
| File | Last Modified |
|---|---|
| clipboard_e0d4b00b708e2951d6398e6cdfb623f3f.png | October 19, 2023 |
| clipboard_ef1a7ddf4d525b9337d7419315b6971c7.png | October 19, 2023 |
| clipboard_efe15ce77c95264adafc242bbbde97c39.png | October 19, 2023 |
| clipboard_eb5ff812270405e9970076e8e4c0ecdc9.png | October 19, 2023 |