Thermo Fisher Scientific
iCAP Pro autopeak will not pass
Issue
The Autopeak will not pass on the iCAP Pro
Environment
  • iCAP Pro
Resolution
Delete the previous autopeak within "View Autopeak Data" and perform the Autopeak again.
  • Was this article helpful?