AN40986 - Environmental Series - Trace contaminant analysis in brine using a Thermo Scientific iCAP 6000 Series Duo ICP
The document details a study on the analysis of trace contaminants in brine using the Thermo Scientific iCAP 6000 Series Duo ICP. Brine samples were tested using 2 methods. Method 1 provided rapid screening with a 5-sec integration time. Method 2 used a longer integration time for better detection limits. Both methods showed high precision and accuracy, confirming that the iCAP 6000 Series is capable of long-term analysis without sensitivity loss and suitable for high-salt matrices analysis.
Description:
Environmental Series - Trace contaminant analysis in brine using a Thermo Scientific iCAP 6000 Series Duo ICP
Environment:
- iCAP 6000 series
- iCAP 7000 series
Attachment(s):
| File | Last Modified |
|---|---|
| AN40986Brine.PDF | April 17, 2023 |