Loaded Blank and Multi Element Test Solutions
Issue
Can I order the Loaded Blank (Setup Solutions) or Multi Element Test (MET) solutions individually?
Environment
- iCAP 6000 Series
- iCAP 7000 Series
- iCAP PRO
Resolution
No, these solutions can only be procured by ordering one of the following kits:
Test Solutions - P/N 842313050711
Contents, Quantity 1 of each:
Setup Solution (Loaded Blank) P/N 430122821411
Mult Element Test Solution P/N 430122821401
PM Kit - P/N BRE0012344
Contents of kit:
Part number Description Quantity
430122821761 2/2 Solenoid Valve 1145BC 24V DC 1
598000000002 Filter Kit, 35Micron, TF9-50 1
430116620622 Ignitor Holder 1
430122821611 Shielding Gasket EPC0622 5 ft
842312051401 Sampling O - Ring Kit 1
430122821401 iCAP 6000 Multi Element Test Solution 1
430122821411 iCAP 6000 Setup Solution 1
842312052621 Concentrated Corrosion Inhibitor Kit 1