Radial POP Interface Kit for Duo
Description:
The dual view purged optical path plasma interfaces of the Thermo Scientific™ iCAP™ 7000 Plus Series ICP-OES enhance long term stability and improve detection limits
in the wavelength region below 180 nm for both axial and radial measurements. This Technical Note: SP-43438-ICP-OES-Purged optical path plasma-iCAP 7000 Plus-SP43438-EN.pdf provides further details on this new interface for the radial view.
The P/N for the Radial POP Interface Kit is: BRE0011655. It contains the following items:
1x POP interface
2x glass window
2x O-ring
1x alignment tool
Environment:
- iCAP 7000 Series
Attachment(s):
| File | Last Modified |
|---|---|
| Radial POP Interface kit.png | January 26, 2023 |
| SP-43438-ICP-OES-Purged optical path plasma-iCAP 7000 Plus-SP43438-EN.pdf | August 02, 2022 |