Sampling O-ring Kit
Description:
The below infor graphic details the size and location of the o-rings found in the sampling O-ring kit - P/N 842312051401. The number of o-rings is also included and is based on the number of o-rings listed in the second column. For example, there are a quantity of 10 of o-ring 'H' used to seal the center tube into the center tube holder. This infographic listing the size of the o-ring can be helpful if you are trying to source only one of the o-rings. If you are going to source one of the o-rings independent of the o-ring kit, make sure you procure one that is composed of Viton.
Environment:
- iCAP 6000 Series
- iCAP 7000 Series