Thermo Fisher Scientific
Z-axis Replacement on Cetac ASX 280, 560 Autosamplers
Issue
Z-axis is not moving correctly. For example, it is not going down all the way.
Environment
  • Cetac ASX-280
  • Cetac ASX-560
Resolution
Replace the Z-axis tubing (P/N BRE0004307) or drive assembly (P/N BRE0004428)
Here is a helpful video on how to replace the Z-axis drive assembly:
Cause
Sometimes the Z-axis tubing can become kinked. Once it becomes kinked it is oftentimes best to replace it.