Thermo Fisher Scientific
Autosampler intermittently loses communication in Windows 11
Issue
Autosampler intermittently loses communication in Windows 11
Environment
  • ASX-560
  • ASX-280
Resolution
Roll back FTDI VCP drivers to 2.12.24.0 (2016) 
Cause
Teledyne Cetac determined that FTDI drviers installed were too new and needed to be downgraded.
  • Was this article helpful?