Cetac ASX-560, ASX-280 home position error and Labbook fails
Issue
After autosampler ASX-560 or ASX-280 installation, it may occur running a labbook, that Qtegra stops when the probe returns to home position from random rack position, the autosampler led in dashboard turns red, showing false communication issue. Error 017: error moving home is an example of an error message that may be present in the log file.
Environment
- iCAP 6000 Series
- iCAP 7000 Series
- ICAPPRO Series
- Qtegra
- ASX 560
- ASX 280
Resolution
See attached file: Cetac ASX-560_280 home position error Rev 2.pdf for more information on the error and this knowledge article for details on how to perform an alignment: Rev 4 - 480221 - Alignment Guide for ASX-280 and ASX-560 Autosamplers
Attachment(s):
File | Last Modified |
---|---|
ASX-280 & ASX-560 Alignment Guide.pdf | August 02, 2022 |
Cetac ASX-560_280 home position error Rev 2.pdf | August 02, 2022 |