Changing the Personality of CETAC Autosampler to Establish Communication with Qtegra
To change the personality of the CETAC Auto Sampler to establish communication with Qtegra, ensure the sampler's communications cable is connected, plugged in, and powered on. Use Sprint Valve Configurator to detect the COM port, record it, then set up the serial port in C-Term using the detected COM port. For ASX 520, set the personality type to 8. Verify control in Qtegra by moving the probe. Communication should now be established.
Description:
The personality should be set by CETAC for every autosampler, which determines the specific device with which it will be communicating. In the event communication does not occur or is not working properly, check or change hte personality for the instrument.
Environment:
- Qtegra
- CETAC Autosampler
Attachment(s):
| File | Last Modified |
|---|---|
| Changing the Personality of CETAC Autosampler Qtegra.pdf | December 09, 2021 |